Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylpropionaldehyde, 97%

Catalog No. p-7026439
Click to view available options
Quantity:
100 g
500 g

CAS: 93-53-8 | C9H10O | 134.178 g/mol

2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

Solubility
Soluble in most fixed oils, propylene glycol. Insoluble in glycerin.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-53-8
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00006973
InChI Key IQVAERDLDAZARL-UHFFFAOYSA-N
Synonym 2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
PubChem CID 7146
IUPAC Name 2-phenylpropanal
SMILES CC(C=O)C1=CC=CC=C1

Specifications

Density 1.01
Boiling Point 92°C to 94°C (12 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.517
Quantity 100 g
Beilstein 4291321
Sensitivity Air Sensitive
Solubility Information Soluble in most fixed oils,propylene glycol. Insoluble in glycerin.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 2-Phenylpropionaldehyde
Show More Show Less
Hazard Category H227-H315-H317-H319-H361f
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P261-P264b-P272-P280-P281-P302+P352-P305+P351+P338-P308+P313-P333+P313-P362-P370+P378q-P501c
EINECSNumber 202-255-5
RTECSNumber CY1460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.