Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylpyridine 98.0+%, TCI America™
SDP

Catalog No. P103925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P103925G 25 g
P10395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P103925G Supplier TCI America Supplier No. P103925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1008-89-5
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.20
MDL Number MFCD00006280,MFCD31699954
InChI Key VQGHOUODWALEFC-UHFFFAOYSA-N
Synonym pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine????
PubChem CID 13887
IUPAC Name 2-phenylpyridine
SMILES C1=CC=C(C=C1)C1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 270°C
Quantity 25 g
Formula Weight 155.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylpyridine
EINECSNumber (5)-3717
RTECSNumber UT7140900
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.