Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylthiazole, 98%

Catalog No. AC394370010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC394370010 1 g
AC394370050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC394370010 Supplier Thermo Scientific Chemicals Supplier No. 394370010
Add to Cart

CAS: 1826-11-5 | C9H7NS | 161.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1826-11-5
Molecular Formula C9H7NS
Molecular Weight (g/mol) 161.23
InChI Key WYKHSBAVLOPISI-UHFFFAOYSA-N
Synonym 2-phenylthiazole, thiazole, 2-phenyl, phenylthiazole, 2-phenyl thiazole, #, ksc174s8f, tpc-i164
PubChem CID 547494
IUPAC Name 2-phenyl-1,3-thiazole
SMILES C1=CC=C(C=C1)C2=NC=CS2

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 161.23
Percent Purity 98%
Chemical Name or Material 2-Phenylthiazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.