Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phthalimidoethanesulfonyl chloride, 95%, Thermo Scientific™

Catalog No. AC430980010
Click to view available options
:
1g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4403-36-5
CAS Min % 94.0
CAS Max % 100.0
Quantity 1g
Assay Percent Range 95%
Molecular Formula C10H8ClNO4S
MDL Number MFCD01861217
Synonym 2-phthalimidoethanesulfonyl chloride, 2-1,3-dioxoisoindolin-2-yl ethanesulfonylchloride, 2-1,3-dioxoisoindolin-2-yl ethanesulfonyl chloride, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethanesulfonyl chloride, 2-phthalimidoethanesulfonylchloride, 2-phthalimido ethanesulfonyl chloride, 2-1,3-dioxoisoindol-2-yl ethanesulfonyl chloride, 2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethane-1-sulfonyl chloride, 2-1,3-dioxo-1,3-dihydro-isoindol-2-yl-ethanesulfonyl chloride, 2-2-chlorosulfonyl ethyl benzo c azoline-1,3-dione
InChI Key HCPVYBCAYPMANM-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCS(=O)(=O)Cl
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)ethanesulfonyl chloride
Molecular Weight (g/mol) 273.687
PubChem CID 308739
Formula Weight 273.7
Percent Purity ≥94.0% (derivatized with diethylamine) (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material 2-Phthalimidoethanesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.