Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Picoline, 98+%

Catalog No. aaa10847ae
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 109-06-8 | C6H7N | 93.129 g/mol

2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, sulfuric acid, acids, metals, iron(II) sulfate and hydrogen peroxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-06-8
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006332
InChI Key BSKHPKMHTQYZBB-UHFFFAOYSA-N
Synonym 2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha
PubChem CID 7975
ChEBI CHEBI:50415
IUPAC Name 2-methylpyridine
SMILES CC1=CC=CC=N1

Specifications

Melting Point -70°C
Density 0.944
Boiling Point 128°C to 129°C
Flash Point 26°C (78°F)
Refractive Index 1.5
Quantity 100 mL
UN Number UN2313
Beilstein 104581
Merck Index 14,7400
Solubility Information Miscible with water.
Formula Weight 93.13
Percent Purity ≥98%
Chemical Name or Material 2-Picoline
Show More Show Less
Hazard Category H226-H302+H332-H311-H319-H335
Hazard Statement GHS H Statement
H226-H302-H312-H332-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES
EINECSNumber 203-643-7
RTECSNumber TJ4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.