Learn More
2-Picoline, 98+%
Description
2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, sulfuric acid, acids, metals, iron(II) sulfate and hydrogen peroxide.
 
            Specifications
Specifications
| Melting Point | -70°C | 
| Density | 0.944 | 
| Boiling Point | 128°C to 129°C | 
| Flash Point | 26°C (78°F) | 
| Refractive Index | 1.5 | 
| Quantity | 100 mL | 
| UN Number | UN2313 | 
| Beilstein | 104581 | 
| Merck Index | 14,7400 | 
| Solubility Information | Miscible with water. | 
| Show More | 
Safety and Handling
| Hazard Category | H226-H302+H332-H311-H319-H335 | 
| Hazard Statement | GHS H Statement H226-H302-H312-H332-H319-H335 Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation. May cause respiratory irritation. | 
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c | 
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES | 
| EINECSNumber | 203-643-7 | 
| RTECSNumber | TJ4900000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                