Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187584 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
10 mL

2-Propanol is a secondary alcohol, which can find a wide number of applications as a solvent.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-63-0
Ignition Point 750°F
Vapor Pressure 44 mmHg (25°C)
Vapor Density 2.1 (vs. air)
Density 0.785 g/mL (at 25°C)
Flash Point 12°C
Physical Form Neat
Linear Formula (CH3)2 CHOH
MDL Number MFCD00011674
Melting Point -89.5°C
Boiling Point 82°C
Synonym sec-Propyl alcohol; IPA; Isopropanol; Isopropyl alcohol
Solubility Completely soluble in water
Type Analytical Standard
Quantity 1 mL
Formula Weight 60.10
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.