Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC), Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications

Specifications
Specifications
pH | Neutral |
Melting Point | -89.5°C |
Color | Colorless |
Density | 0.785g/mL at 25°C |
Boiling Point | 77°C |
Quantity | 2.5 L |
Explosion Limit | 0.02 |
Flash Point | 12°C (53.6°F) |
Ignition Point | 750°F |
Odor | Alcohol-like |
Show More |
Safety and Handling
Hazard Statement | P210-P261-P305 + P351 + P338 |
DOTInformation | Class 3 - PG 2 - Isopropanol |
RTECSNumber | 200-661-7 |
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction