Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol (USP/EP/BP/JP), Fisher Chemical™

Catalog No. p-217406 Shop All Fisherbrand Products
Click to view available options
Quantity:
20 L
Packaging:
Steel Pail
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Propanol (USP/EP/BP/JP)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Absorbance Pass Test (BP), Pass Test (BP/EP), Pass Test (EP)
Color Colorless
Acidity Pass Test (JP), Pass Test (JP/USP), Pass Test (USP)
Appearance Clear and complete (EP)
Density 0.785g/cm³
Boiling Point 81°C
Quantity 20 L
Identification Pass Test (BP), Pass Test (BP/EP/JP/USP), Pass Test (EP), Pass Test (JP), Pass Test (USP)
Packaging Steel Pail
Physical Form Liquid
Refractive Index 1.376 to 1.378 (at 20°C) (USP)
Residue after Evaporation 0.005% max. (JP)
Specific Gravity 0.783 to 0.787 (USP), 0.785 to 0.788 (JP), 0.785 to 0.788 (JP), 0.783 to 0.787 (USP)
Linear Formula (CH3)2CHOH
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPa/s at 20°C
Water 0.5% Max. (BP/EP), 0.75% Max. (JP), 0.5% max. (EP), 0.75% max. (JP)
Formula Weight 60.1
Percent Purity ≥99.0% (USP)
Grade BP/EP/FCC/JP/NF/USP
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product.
 
Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator and end-user to qualify, test and/or validate each product for its intended use as an ingredient in the end product and in accordance with applicable regulations (including without limitation WHO-recommended Hand Rub Formulations (Hand Sanitizers) and/or FDA Policy for Preparation of Certain Alcohol-based Hand Sanitizer Products, as updated). Thermo Fisher Scientific expressly disclaims all non-specified warranties, expressed or implied, including the implied warranties of merchantability and fitness for a particular use.
 

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.