Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propionylfuran, 97%

Catalog No. p-7029306
Click to view available options
Quantity:
5 g
25 g

CAS: 3194-15-8 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3194-15-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00020936
InChI Key HCPORNAVHSWTOJ-UHFFFAOYSA-N
Synonym 2-propionylfuran, 1-2-furyl propan-1-one, 1-furan-2-yl propan-1-one, 1-propanone, 1-2-furanyl, 2-furyl ethyl ketone, ethyl 2-furyl ketone, furyl ethyl ketone, 1-propanone, 1-2-furyl, ethyl furyl ketone, 2-propanoylfuran
PubChem CID 76662
IUPAC Name 1-(furan-2-yl)propan-1-one
SMILES CCC(=O)C1=CC=CO1

Specifications

Melting Point 26°C to 30°C
Boiling Point 53°C to 55°C (1 mmHg)
Odor Characteristic
Quantity 5 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Propionylfuran
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 221-693-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.