Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylcyclohexanol (cis- and trans- mixture) 90.0+%, TCI America™
SDP

Catalog No. P077025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P077025G 25 g
P07705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P077025G Supplier TCI America Supplier No. P077025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90676-25-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00019331
InChI Key VZBNUCDUQJCIDP-UHFFFAOYNA-N
PubChem CID 138614
IUPAC Name 2-propylcyclohexan-1-ol
SMILES CCCC1CCCCC1O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 142.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylcyclohexanol (cis- and trans- mixture)
EINECSNumber (3)-2301
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.