Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylfuran 98.0+%, TCI America™
SDP

Catalog No. P148825G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4229-91-8
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00047070
InChI Key CPLJMYOQYRCCBY-UHFFFAOYSA-N
Synonym furan, 2-propyl, 2-n-propylfuran, 2-n-propyl furan, furan, propyl, 2-propyl-furan, furan, alpha-propyl, acmc-1ctud, 2-propyl furan, furan, .alpha.-propyl, cpljmyoqyrccby-uhfffaoysa
PubChem CID 77907
IUPAC Name 2-propylfuran
SMILES CCCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
UN Number 1993
Formula Weight 110.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylfuran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.