Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylpentanoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
valproic acid,dipropylacetic acid,depakene,depakine,2-propylvaleric acid,ergenyl,di-n-propylacetic acid,valproate,mylproin,pentanoic acid, 2-propyl
3121
CHEBI:39867
2-propylpentanoic acid
CCCC(CCC)C(=O)O
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
valproic acid,dipropylacetic acid,depakene,depakine,2-propylvaleric acid,ergenyl,di-n-propylacetic acid,valproate,mylproin,pentanoic acid, 2-propyl
3121
CHEBI:39867
2-propylpentanoic acid
CCCC(CCC)C(=O)O
Specific Gravity 0.92
Quantity 25g
Refractive Index 1.4240 to 1.4260
Boiling Point 220.0°C
Density 0.9200g/mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Flash Point 111°C
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula (CH3CH2CH2)2CHCO2H
Beilstein 02, 350
Merck Index 15, 10099
Solubility Information Solubility in water: slightly soluble
Formula Weight 144.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Propylpentanoic acid, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May damage the unborn child.
Suspected of damaging fertility.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

EINECSNumber : 202-777-3

RTECSNumber : YV7875000

TSCA : TSCA