Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Propylpentanoic acid, 99%, ACROS Organics™
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Refractive Index | 1.4240 to 1.4260 |
Specific Gravity | 0.92 |
Infrared Spectrum | Authentic |
Boiling Point | 220.0°C |
Quantity | 100g |
Density | 0.9200g/mL |
Flash Point | 111°C |
Color | Colorless to Yellow |
CAS Min % | 98.5 |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May damage the unborn child.
Suspected of damaging fertility.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
EINECSNumber : 202-777-3
RTECSNumber : YV7875000
TSCA : TSCA