Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dibenzoyl peroxide, 75%, remainder water
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators

Specifications
Specifications
Melting Point | 104.0°C to 106.0°C |
Color | White |
Packaging | Plastic bottle |
Linear Formula | (C6H5CO)2O2 |
Quantity | 1 kg |
Beilstein | 09, 179 |
Fieser | 01,196; 04,122; 05,182; 06,160; 07,89; 12,157; 15,146; 17,367 |
Merck Index | 15, 1119 |
Solubility Information | Solubility in water: insoluble. Other solubilities: sparingly soluble in alcohol,soluble in benzene,chloroform,ether and acetone,1g/40 mL carbon disulfide,1g/40 mL olive oil |
Formula Weight | 242.23 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause an allergic skin reaction. Causes serious eye irritation. Heating may cause a fire. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep/Store away from clothing/ combustible materials. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash |
EINECSNumber | 202-327-6 |
RTECSNumber | DM8575000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction