Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyrazinecarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
98-97-5
C5H4N2O2
124.1
MFCD00006130
NIPZZXUFJPQHNH-UHFFFAOYSA-N
2-pyrazinecarboxylic acid,pyrazinoic acid,pyrazinecarboxylic acid,pyrazinic acid,2-carboxypyrazine,paradiazinecarboxylic acid,pyrazinemonocarboxylic acid,2-pyrazinoic acid,2-pyrazine carboxylic acid,piazinecarboxylic acid
1047
CHEBI:71311
pyrazine-2-carboxylic acid
C1=CN=C(C=N1)C(=O)O
98-97-5
C5H4N2O2
124.1
MFCD00006130
NIPZZXUFJPQHNH-UHFFFAOYSA-N
2-pyrazinecarboxylic acid,pyrazinoic acid,pyrazinecarboxylic acid,pyrazinic acid,2-carboxypyrazine,paradiazinecarboxylic acid,pyrazinemonocarboxylic acid,2-pyrazinoic acid,2-pyrazine carboxylic acid,piazinecarboxylic acid
1047
CHEBI:71311
pyrazine-2-carboxylic acid
C1=CN=C(C=N1)C(=O)O
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 222.0°C to 225.0°C
Quantity 25g
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 25, 125
Merck Index 15, 8070
Solubility Information Solubility in water: soluble in cold water. Other solubilities: 1g/120g absolubte ethanol (25 c),practically insoluble in ether,chloroform and,benzene
Formula Weight 124.1
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2-Pyrazinecarboxylic acid, 99%
Show More Show Less

EINECSNumber : 202-718-1

TSCA : TSCA