Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-sec-Butyl-3-methoxypyrazine, 99%, Thermo Scientific™

Catalog No. AC310460010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 24168-70-5
Molecular Formula C9H14N2O
Molecular Weight (g/mol) 166.224
MDL Number MFCD00006136
InChI Key QMQDJVIJVPEQHE-UHFFFAOYSA-N
Synonym 2-sec-butyl-3-methoxypyrazine, 2-methoxy-3-sec-butylpyrazine, 2-methoxy-3-1-methylpropyl pyrazine, pyrazine, 2-methoxy-3-1-methylpropyl, 2-methoxy-3-sec-butyl pyrazine, 3-sec-butyl-2-methoxypyrazine, pyrazine, 2-methoxy, 3-sec-butyl, 2-methoxy-3-1-methyl-propyl pyrazine
PubChem CID 520098
IUPAC Name 2-butan-2-yl-3-methoxypyrazine
SMILES CCC(C)C1=NC=CN=C1OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9900g/mL
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Refractive Index 1.4920 to 1.4960
Quantity 1 g
Spectrophotometry 98.5% min.
Formula Weight 166.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-sec-Butyl-3-methoxypyrazine, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.