Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-TEDC, Tocris Bioscience™

Catalog No. p-4783174
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-, 12-, 15-Lipoxygenase inhibitor

Potent inhibitor of 5-, 12-, and 15-lipoxygenase (IC50 values are 0.09, 0.013 and 0.5 μM respectively).

Chemical Identifiers

CAS 132465-10-2
Molecular Formula C16H13NO4S
Molecular Weight (g/mol) 315.343
InChI Key KCGLTUBCAGZLHP-XYOKQWHBSA-N
Synonym 2-tedc, chembl37081, tocris-0645, 2-thiophen-2-yl ethyl 2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, e-2-cyano-3-3,4-dihydroxyphenyl acrylic acid 2-2-thienyl ethyl ester, 2-cyano-3-3,4-dihydroxy-phenyl-acrylic acid 2-thiophen-2-yl-ethyl ester, 2-cyano-3-3,4-dihydroxyphenyl propenoic acid 2-2-thienyl ethyl ester, 2-thiophen-2-ylethyl e-2-cyano-3-3,4-dihydroxyphenyl prop-2-enoate, 2-propenoic acid,2-cyano-3-3,4-dihydroxyphenyl-,2-2-thienyl ethyl ester, e-9ci
PubChem CID 6069552
IUPAC Name 2-thiophen-2-ylethyl (E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES C1=CSC(=C1)CCOC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Quantity 10 mg
Formula Weight 315.34
Percent Purity >99%
Chemical Name or Material 2-TEDC
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.