Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Butyl-6-methylaniline 98.0+%, TCI America™
SDP

Catalog No. M07635ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL

Chemical Identifiers

CAS 13117-94-7
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00025208
InChI Key GWIUAENVORNODL-UHFFFAOYSA-N
Synonym 6-tert-Butyl-o-toluidine
PubChem CID 83155
IUPAC Name 2-tert-butyl-6-methylaniline
SMILES CC1=C(C(=CC=C1)C(C)(C)C)N

Specifications

Boiling Point 250°C
Quantity 5 mL
Formula Weight 163.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-tert-Butyl-6-methylaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.