Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Butylanthraquinone 95%, Thermo Scientific™

Catalog No. AC148565000
Click to view available options
Quantity:
100 g
500 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-47-9
Molecular Formula C18H16O2
Molecular Weight (g/mol) 264.32
MDL Number MFCD00001232
InChI Key YTPSFXZMJKMUJE-UHFFFAOYSA-N
Synonym 2-tert-butylanthraquinone, 9,10-anthracenedione, 2-1,1-dimethylethyl, 2-tert-butyl-9,10-anthraquinone, 2-tert-butyl anthracene-9,10-dione, anthraquinone, 2-tert-butyl, beta-tert-butylanthraquinone, 2-t-butylanthraquinone, .beta.-tert-butylanthraquinone, acmc-209puu, 2-tert-butyl-anthrachinon
PubChem CID 66532
IUPAC Name 2-tert-butyl-9,10-dihydroanthracene-9,10-dione
SMILES CC(C)(C)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 98°C to 103°C
Color Yellow
Infrared Spectrum Authentic
Quantity 500 g
Formula Weight 264.32
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material 2-tert-Butylanthraquinone, 95%
EINECSNumber 201-531-2
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.