Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich 2-Tert-Butylimino-2-Diethylamino-1,3-Dimethylperhydro-1,3,2-Diazaphosphorine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Description
2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine (BEMP) is a phosphazene base. BEMP participates in the mild base catalyzed nucleophilic ring opening of N-sulfonyl aziridines. BEMP supported on polystyrene (PS-BEMP) has been reported as an efficient catalyst for the ring-opening of epoxides with phenols. BEMP is about 2000 times more basic and also much more sterically hindered than DBU (l,8-diazabicyclo[5.4.0]undecene).Phosphazene Bases: a Family of Extremely Strong, Non-Ionic, Non-Charged Nitrogen-Bases
Specifications
Specifications
| CAS | 98015-45-3 |
| Density | 0.948 g/mL at 25°C |
| Boiling Point | 74°C (0.03 mmHg, lit.) |
| Molecular Formula | C13H31N4P |
| Refractive Index | n20/D 1.477; n20/D 1.477 |
| Linear Formula | C13H31N4P |
| MDL Number | MFCD00043143 |
| Quantity | 1 mL |
| Synonym | BEMP |
| Molecular Weight (g/mol) | 274.39 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction