Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Tetralone, 99%

Catalog No. aal0105306
Click to view available options
Quantity:
5 g
25 g

CAS: 530-93-8 | C10H10O | 146.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-93-8
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00001727
InChI Key KCKZIWSINLBROE-UHFFFAOYSA-N
Synonym 2-tetralone, beta-tetralone, 1,2,3,4-tetrahydro-2-naphthalenone, 3,4-dihydronaphthalen-2 1h-one, 1,2,3,4-tetrahydronaphthalen-2-one, 2 1h-naphthalenone, 3,4-dihydro, .beta.-tetralone, tetralin-2-one, 3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-2-oxonaphthalene
PubChem CID 68266
IUPAC Name 3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=CC=CC=C2CC1=O

Specifications

Melting Point 18°C
Density 1.106
Boiling Point 240°C
Flash Point >110°C (230°F)
Refractive Index 1.56
Quantity 5 g
Beilstein 509386
Sensitivity Air sensitive
Formula Weight 146.19
Percent Purity 99%
Chemical Name or Material 2-Tetralone
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-498-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.