Learn More
2-Thionaphthol, 98%
Description
2-Thionaphthol was used in the preparation of cholesterol monolayer and multilayer Langmuir- Blodgett (LB) films. The magnetic resonance shift for a self-assembled monolayer of 2-naphthalenethiol was studied that suggested considerable promise in flexible and transparent photonic devices for biological and chemical sensing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Thionaphthol was used in the preparation of cholesterol monolayer and multilayer Langmuir- Blodgett (LB) films. The magnetic resonance shift for a self-assembled monolayer of 2-naphthalenethiol was studied that suggested considerable promise in flexible and transparent photonic devices for biological and chemical sensing.
Solubility
Soluble in alcohol, dipropylene glycol. Soluble in water 34.4 mg/L @ 25°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 79°C to 83°C |
| Density | 1.55 |
| Boiling Point | 286°C |
| Quantity | 10 g |
| Beilstein | 636389 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,6379 |
| Solubility Information | Soluble in alcohol,dipropylene glycol. Soluble in water 34.4mg/L @ 25°C. |
| Formula Weight | 160.24 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 202-082-5 |
| RTECSNumber | QK3930000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.