Learn More
2-Thiopheneacetic acid, 98%
Description
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Thiopheneacetic acid is used in the preparation of rosette-like nanoscale gold materials. Further, it is used as a pharmaceutical intermediate for cefaloridine and cefalotin sodium.
Solubility
Soluble in water, ethanol, ether and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents and strong bases.

Specifications
Specifications
Melting Point | 61°C to 65°C |
Boiling Point | 160°C (22 mmHg) |
Quantity | 500 g |
UN Number | UN3261 |
Beilstein | 114551 |
Solubility Information | Soluble in water,ethanol,ether and carbon tetrachloride. |
Formula Weight | 142.18 |
Percent Purity | 98% |
Chemical Name or Material | 2-Thiopheneacetic acid |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 217-639-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.