Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T177225G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Specifications

CAS 6165-68-0
Color White-Yellow
Molecular Formula C4H5BO2S
Quantity 25 g
MDL Number MFCD00151850
Synonym thiophene-2-boronic acid, 2-thiopheneboronic acid, 2-thienylboronic acid, thienylboronic acid, 2-thiopheneboronicacid
InChI Key ARYHTUPFQTUBBG-UHFFFAOYSA-N
SMILES B(C1=CC=CS1)(O)O
IUPAC Name thiophen-2-ylboronic acid
Molecular Weight (g/mol) 127.952
PubChem CID 2733960
Formula Weight 127.95
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.