Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenecarboxylic acid, 99%

Catalog No. p-3758816
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

CAS: 527-72-0 | C5H4O2S | 128.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-72-0
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.15
MDL Number MFCD00005437
InChI Key QERYCTSHXKAMIS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid
PubChem CID 10700
ChEBI CHEBI:71241
IUPAC Name thiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=CS1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 126.0°C to 130.0°C
Boiling Point 260.0°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18,289
Merck Index 15,9507
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in alcohol and ether, moderately soluble in chcl3, slightly soluble in petroleum ether
Formula Weight 128.15
Percent Purity 99%
Chemical Name or Material 2-Thiophenecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 208-423-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.