Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneethanol 98.0+%, TCI America™
SDP

Catalog No. T1021250ML
Click to view available options
Quantity:
25 mL
250 mL

Chemical Identifiers

CAS 5402-55-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00005462
InChI Key VMJOFTHFJMLIKL-UHFFFAOYSA-N
Synonym 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
PubChem CID 79400
IUPAC Name 2-(thiophen-2-yl)ethan-1-ol
SMILES OCCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 250 mL
Formula Weight 128.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Thiopheneethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.