Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenemethylamine, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
27757-85-3
C5H7NS
113.18
MFCD00005460
FKKJJPMGAWGYPN-UHFFFAOYSA-N
2-thiophenemethylamine,2-thienylmethylamine,2-thiophenemethanamine,2-aminomethyl thiophene,thiophene-2-methylamine,thiophenemethanamine,2-aminomethylthiophene,2-thenylamine,2-thienylmethyl amine,1-thiophen-2-yl methanamine
34005
thiophen-2-ylmethanamine
C1=CSC(=C1)CN
27757-85-3
C5H7NS
113.18
MFCD00005460
FKKJJPMGAWGYPN-UHFFFAOYSA-N
2-thiophenemethylamine,2-thienylmethylamine,2-thiophenemethanamine,2-aminomethyl thiophene,thiophene-2-methylamine,thiophenemethanamine,2-aminomethylthiophene,2-thenylamine,2-thienylmethyl amine,1-thiophen-2-yl methanamine
34005
thiophen-2-ylmethanamine
C1=CSC(=C1)CN
Packaging Glass bottle
Specific Gravity 1.1
Refractive Index 1.5650 to 1.5680
Density 1.1000g/mL
Flash Point 73°C
Quantity 25g
Infrared Spectrum Authentic
Color Colorless to Yellow
Boiling Point 95.0°C to 99.0°C (28.0mmHg)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Beilstein 18, IV, 7096
Formula Weight 113.18
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-Thiophenemethylamine, 90-96%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 248-639-6

TSCA : TSCA