Learn More
2-Thiophenemethylamine, 94%
Description
2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.
Solubility
Easily soluble in ethanol and Chloroform.
Notes
Air Sensitive. Store under dry inert gas. Store away from air.

Specifications
Specifications
Density | 1.1 |
Boiling Point | 85°C to 88°C (14 mmHg) |
Flash Point | 73°C (163°F) |
Refractive Index | 1.567 |
Quantity | 5 g |
UN Number | UN2735 |
Beilstein | 106296 |
Sensitivity | Air Sensitive |
Solubility Information | Easily soluble in ethanol and Chloroform. |
Formula Weight | 113.19 |
Show More |
Safety and Handling
Hazard Category | H227-H314-H335 |
Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
Precautionary Statement | P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
EINECSNumber | 248-639-6 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.