Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thioxotetrahydro-1,3-oxazole, 98%, Thermo Scientific™

Catalog No. ac304135000
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle

Chemical Identifiers

CAS 5840-81-3
Molecular Formula C3H5NOS
Molecular Weight (g/mol) 103.14
MDL Number MFCD00125763
InChI Key UMURLIQHQSKULR-UHFFFAOYSA-N
Synonym oxazolidine-2-thione, 2-oxazolidinethione, oxazolidinethione, 2-thiooxazolidone, 2-mercapto-2-oxazoline, 2-thioxotetrahydro-1,3-oxazole, unii-6smj3cuw4c, 1,3-oxazolane-2-thione, 6smj3cuw4c, 4,5-dihydro-1,3-oxazole-2-thiol
PubChem CID 2733223
IUPAC Name 1,3-oxazolidine-2-thione
SMILES C1COC(=S)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Boiling Point 85.0°C to 91.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 103.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Thioxotetrahydro-1, 3-oxazole, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.