Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Tri-n-butylstannyl)furan, 97%

Catalog No. AAH55259AE
Click to view available options
Quantity:
5 mL
25 mL
100 mL

Catalog No. AAH55259AE

Supplier: Thermo Scientific Chemicals H55259AE

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CAS: 118486-94-5 | C16H30OSn | 357.125 g/mol

It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Store at room temperature. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118486-94-5
Molecular Formula C16H30OSn
Molecular Weight (g/mol) 357.125
MDL Number MFCD00192512
InChI Key SANWDQJIWZEKOD-UHFFFAOYSA-N
Synonym 2-tributylstannyl furan, tributyl furan-2-yl stannane, 2-tributylstannylfuran, tributyl 2-furyl tin, tributyl 2-furyl stannane, 2-tri-n-butylstannyl furan, stannane,tributyl-2-furanyl, stannane, tributyl-2-furanyl, 2-furyltributyltin, tributylstannylfuran
PubChem CID 4393688
IUPAC Name tributyl(furan-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CO1

Specifications

Color Colorless to Yellow
Density 1.134 g/mL
Boiling Point 90°C (0.1 mmHg)
Refractive Index 1.494
Quantity 100 mL
UN Number UN2788
Formula Weight 357.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)furan
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.