Alfa Aesar™ 2-(Tri-n-butylstannyl)furan, 97%

Manufacturer: Alfa Aesar™ H55259AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH55259AA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 118486-94-5
    Formula Weight 357.1
    MDL Number MFCD00192512
    UN Number UN2788
    Chemical Name or Material 2-(Tri-n-butylstannyl)furan

    Specifications

    Formula Weight 357.1
    Health Hazard 3 GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Refractive Index 1.4940
    MDL Number MFCD00192512
    CAS 118486-94-5
    UN Number UN2788
    Health Hazard 1 Danger
    Density 1.134g/mL
    Boiling Point 90°C (0.1mmHg)
    Physical Form Liquid
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Color Colorless to yellow
    Quantity 5mL
    Chemical Name or Material 2-(Tri-n-butylstannyl)furan
    Assay 97%

    Description

    Applications
    It is an agrochemical, organic and pharmaceutical intermediates.

    Notes
    Store at room temperature. Light sensitive.

    GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit