Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Tributylstannyl)Pyrimidine

Catalog No. 7211745G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
1 g
5 g
Danger

Catalog No. 72-117-45G

Supplier: Sigma Aldrich 7211745G

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Tributylstannyl)pyrimidine is an organotin compound used in Stille coupling reaction. This can be used as one of the precursors in the synthesis of 2-aminopyridine oxazolidinones as potent and selective tankyrase (TNKS) inhibitors and the synthesis of canagliflozin, a novel inhibitor for sodium-dependent glucose cotransporter.It can also be used in the preparation of various (2-pyrimidyl)silanes.

Specifications

CAS 153435-63-3
Density 1.164 g/mL at 25°C
Molecular Formula C16H30N2Sn
Refractive Index n20/D 1.512
Linear Formula C16H30N2Sn
MDL Number MFCD01319085
Quantity 5 g
Synonym 2-Tributylstannanylpyrimidine; Tributyl (2-pyrimidyl)stannane
Molecular Weight (g/mol) 369.13 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.