Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trichloroacetyl)pyrrole 98.0+%, TCI America™
SDP

Catalog No. T161925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T161925G 25 g
T16195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T161925G Supplier TCI America Supplier No. T161925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35302-72-8
Molecular Formula C6H4Cl3NO
Molecular Weight (g/mol) 212.454
MDL Number MFCD00128757
InChI Key BBFDGMDENAEMKF-UHFFFAOYSA-N
Synonym 2-trichloroacetyl pyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one, 2-trichloroacetyl-1h-pyrrole, 2-trichloroacetylpyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone, ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl, 2,2,2-trichloro-1-pyrrol-2-ylethan-1-one, trichloroacetyl pyrrole
PubChem CID 321487
IUPAC Name 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
SMILES C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl

Specifications

Melting Point 76°C
Quantity 25 g
UN Number 1759
Formula Weight 212.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trichloroacetyl)pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.