Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)benzimidazole, 98%, Thermo Scientific™

Catalog No. AC297890250
Click to view available options
:
25g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 312-73-2
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H5F3N2
Quantity 25g
MDL Number MFCD00022675
Synonym 2-trifluoromethyl benzimidazole, 2-trifluoromethyl-1h-benzo d imidazole, 2-trifluoromethyl-1h-benzimidazole, 2-trifluoromethyl-1h-1,3-benzodiazole, 2-trifluoromethylbenzimidazole, 2-trifluoromethyl-1h-benzoimidazole, 1h-benzimidazole, 2-trifluoromethyl, 2-cf3 benzimidazole, chembl55641
InChI Key MXFMPTXDHSDMTI-UHFFFAOYSA-N
SMILES FC(F)(F)C1=NC2=CC=CC=C2N1
IUPAC Name 2-(trifluoromethyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol) 186.14
PubChem CID 67560
Formula Weight 186.14
Percent Purity ≥97.5%
Chemical Name or Material 2-(Trifluoromethyl)benzimidazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide
RTECSNumber DE1576000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.