Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. C100425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C100425G 25 g
C10045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C100425G Supplier TCI America Supplier No. C100425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 447-60-9
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001791
InChI Key SOZGHDCEWOLLHV-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
PubChem CID 67972
IUPAC Name 2-(trifluoromethyl)benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C(F)(F)F

Specifications

Color Yellow
Boiling Point 206°C
Quantity 25 g
UN Number 3276
Formula Weight 171.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.