Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193343
Click to view available options
Quantity:
5 g
25 g

Specifications

CAS 1423-27-4
Color White-Yellow
Molecular Formula C7H6BF3O2
Quantity 5 g
MDL Number MFCD00236059
Synonym 2-trifluoromethyl phenylboronic acid, 2-trifluoromethylphenylboronic acid, 2-trifluoromethyl phenyl boronic acid, 2-trifluoromethyl benzeneboronic acid, 2-trifluoromethyl phenyl boranediol
InChI Key JNSBEPKGFVENFS-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1C(F)(F)F)(O)O
IUPAC Name [2-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 189.928
PubChem CID 2734387
Formula Weight 189.93
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.