Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-(Trifluoromethyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog No. p-7031444
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131922-05-9
Molecular Formula C5H9F3N2
Molecular Weight (g/mol) 154.136
MDL Number MFCD07373382
InChI Key YNEJOOCZWXGXDU-UHFFFAOYSA-N
Synonym 2-trifluoromethyl piperazine, 2-trifluoromethyl-piperazine, +/--2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl, -2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl-9ci, acmc-20aolo, 2trifluoromethylpiperazine, m-trifluoromethylpiperazine
PubChem CID 18519725
IUPAC Name 2-(trifluoromethyl)piperazine
SMILES C1CNC(CN1)C(F)(F)F

Specifications

Melting Point 94°C to 97°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 154.13
Percent Purity 97%
Chemical Name or Material (+/-)-2-(Trifluoromethyl)piperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.