Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Trimethylsilyloxy-3-butyne, 97%, Thermo Scientific™

Catalog No. p-7035750
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17869-76-0
Molecular Formula C7H14OSi
Molecular Weight (g/mol) 142.27
MDL Number MFCD00190215
InChI Key RMKACVRKUKSWNQ-UHFFFAOYNA-N
Synonym 2-trimethylsilyloxy-3-butyne, 3-trimethylsiloxy-1-butyne, 3-trimethylsilyloxy-1-butyne, but-3-yn-2-yloxy trimethyl silane, silane, trimethyl 1-methyl-2-propynyl oxy, but-3-yn-2-yloxy trimethylsilane, 2-trimethylsilyloxybut-3-yne, trimethyl 1-methyl-2-propynyl oxy silane, 3-butyn-2-ol, tms derivative
PubChem CID 28817
IUPAC Name (but-3-yn-2-yloxy)trimethylsilane
SMILES CC(O[Si](C)(C)C)C#C

Specifications

Boiling Point 116°C
Quantity 5 g
UN Number UN1993
Flash Point 6°C (43°F)
Formula Weight 142.27
Percent Purity 97%
Refractive Index 1.405
Sensitivity Moisture sensitive
Density 0.818
Chemical Name or Material 2-Trimethylsilyloxy-3-butyne
Hazard Category H224-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.