Alfa Aesar™ 2-(Triphenylphosphonio)propiophenone bromide, 98+%

Manufacturer: Alfa Aesar™ B2484506

Quantity 5g

 View more versions of this product

Catalog No. AAB2484506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2689-63-6
    Molecular Formula C27H24BrOP
    Formula Weight 475.37
    MDL Number MFCD00050279
    Beilstein 4116852
    Synonym (1-Benzoylethyl)triphenylphosphonium bromide; (alpha-Methylphenacyl)triphenylphosphonium bromide
    Chemical Name or Material 2-(Triphenylphosphonio)propiophenone bromide

    Specifications

    Formula Weight 475.37
    Molecular Formula C27H24BrOP
    MDL Number MFCD00050279
    CAS 2689-63-6
    Synonym (1-Benzoylethyl)triphenylphosphonium bromide; (alpha-Methylphenacyl)triphenylphosphonium bromide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 244° to 246°C
    Odor Odorless
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Beilstein 4116852
    Chemical Name or Material 2-(Triphenylphosphonio)propiophenone bromide
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit