Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Vinylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Specifications
Specifications
Chemical Name or Material | 2-Vinylphenylboronic Acid (contains varying amounts of Anhydride) |
CAS | 15016-42-9 |
Melting Point | 109°C |
Color | White-Yellow |
Molecular Formula | C8H9BO2 |
Quantity | 1 g |
MDL Number | MFCD01074659 |
Synonym | 2-vinylphenylboronic acid, 2-vinylbenzeneboronic acid, 2-vinylphenyl boronic acid, 2-ethenylphenyl boronic acid, 2-boronostyrene, styrene-2-boronic acid, boronic acid, ethenylphenyl, 2-vinylphenylboronicacid, acmc-1bt6g, 2-vinyl-phenylboronic acid |
InChI Key | QHFAXRHEKNHTDH-UHFFFAOYSA-N |
SMILES | B(C1=CC=CC=C1C=C)(O)O |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction