Learn More
BINDER™ RS 485 Interface (Two Wire)
Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series KBF P
Description
- Additional serial interface can be used parallel to Ethernet
- For Multi Management Software APT-COM™
Specifications
Specifications
Description | RS 485 interface, 2-wire - Additional serial interface can be used parallel to Ethernet, for Multi Management Software APT-COM™ |
For Use With (Equipment) | BINDER Chambers Series KBF P |
Material | Plastic |
Type | RS 485 Interface |