Learn More
Invitrogen™ Thrombospondin 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149867
Description
Thrombospondin 4 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Thrombospondin-4 (THBS4) belongs to the thrombospondin protein family, a family of adhesive calcium-binding glycoproteins that mediate cell-to-cell and cell-to-matrix interactions. Studies suggest that this protein may be involved in local signaling in the developing and adult nervous system. THBS4 is widely expressed and is abundant in atherosclerotic lesions and in areas prone to the development of lesions. It has been suggested to play an important role in the recruitment of macrophages and the local regulation of inflammation associated with atherogenesis. THBS4 has also been suggested as a putative tumor-suppressor protein in colorectal cancer.
Specifications
| Thrombospondin 4 | |
| Polyclonal | |
| Unconjugated | |
| THBS4 | |
| THBS4; thrombospondin 4; thrombospondin-4; TSP4; TSP-4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 21828, 29220, 7060 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P35443, P49744, Q9Z1T2 | |
| THBS4 | |
| Recombinant Thrombospondin 4 (THBS4) (Gly662-Leu923). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.