Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2022-09-05 00:00:00 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc SEPT501AP

Description
2022-09-05 00:00:00 Polyclonal antibody specifically detects 2022-09-05 00:00:00 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
2022-09-05 00:00:00 | |
Polyclonal | |
Unconjugated | |
SEPTIN5 | |
5-Sep; CDCREL; CDCREL1; CDCREL-1; CDCrel-1A; cell division control related protein 1; cell division control-related protein 1; H5; HCDCREL-1; peanut-like 1; peanut-like 1 homolog; peanut-like protein 1; platelet glycoprotein Ib beta chain; Pnutl1; Sept5; septin 5; SEPTIN5; septin-5 | |
Synthetic peptide taken within amino acid region 300-350 on SEPT5 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q99719, Q9JJM9, Q9Z2Q6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116728, 18951, 5413 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction