Learn More
Heidolph™ Pilot Process Standard Jacketed Vessel Kits
SDP
Catalog No. 13890023 Shop All Heidolph North America Products
Click to view available options
Type:
Single Jacketed
Catalog No. 13-890-023
Supplier: Heidolph™ 015908016
Only null left
Add to Cart
Add to Cart
Request a Quote
Heidolph™ Pilot Process Standard Jacketed Vessel Kits for use with Reactor-Ready Pilot Lab Reactor
  • Vessels have 1.25 to 1 ratio of internal height to diameter to mimic plant scale reactors
  • DN200 vessel flange
Specifications
Includes Process Vessel, Turbine Stirrer Paddle, PTFE Pt100 Temperature Probe with Lemo Plug, PTFE Temperature Probe Adapter
Type Single Jacketed
Capacity (Metric) 20 L
For Use With (Equipment) Reactor-Ready Pilot Lab Reactor
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.