Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

20S Proteasome Antibody (863425) [DyLight 405], Novus Biologicals™
SDP

Catalog No. FAB95541E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB95541E 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB95541E Supplier Novus Biologicals Supplier No. FAB95541E
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

20S Proteasome Monoclonal antibody specifically detects 20S Proteasome in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen 20S Proteasome
Applications Immunohistochemistry
Classification Monoclonal
Clone 863425
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Human 20S Proteasome purified from human erthrocytes
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.