Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human CNTF Recombinant Protein, PeproTech®

Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
Description
The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.

Specifications
Specifications
Accession Number | P26441 |
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | Protein with no preservative |
Gene ID (Entrez) | 1270 |
Molecular Weight (g/mol) | 22.8 kDa |
Name | CNTF |
Quantity | 20 μg |
Storage Requirements | -20°C |
Regulatory Status | RUO |
Endotoxin Concentration | <1 EU/μg |
Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction