Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Full NBR Coated Synthetic Knit Gloves with Nitrile Coated Palm

Catalog No. 17200332
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

FlexTech™ Full NBR Coated Synthetic Knit Gloves with Nitrile Coated Palm are lightweight, synthetic knit shells that are double coated with a layer of nitrile rubber (NBR) for water-resistance, and an additional layer of nitrile coating on the palm and fingers for enhanced grip capabilities.

  • Lightweight, synthetic knit shell with double coating
  • Sandy nitrile coated palm and fingertips enhances grip
  • Full NBR coating provides an additional layer of protection against liquids
  • Gloves offer a balance of comfort and dexterity

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Palm, Fingers
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Nitrile Rubber
Product Type Dip-coated Gloves
Size 2X-Large
For Use With (Application) General Assembly, Small Parts Handling, Painting, and Manufacturing
Glove Cuff Color Gray
Glove Cuff Style Knit
Series Y9289
Yarn Gauge 13
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.