Learn More
Poltex Double Bulk Bin With Lid, 4 in. Wide Comp
SDP
Catalog No. 21377539
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Tape Mount
Wall Mount or Countertop
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-539 Tape Mount
21-377-538 Magnetic Mount
21-377-540 Wall Mount or Countertop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-377-539 Supplier Poltex Supplier No. BLKBNLDDBL4T
Add to Cart
Add to Cart
Simple 4 in. two-compartment bulk bin with top opening with lid for filling and front openings for dispensing.
Simple two-compartment bulk bin with lid
Durable and cleanable PETG construction
4 in. wide
Choose from:
  • Wall or countertop mount with rubber bumpons on base
  • VHB tape mount (very high bond tape included)
  • Magnet mount (powerful magnets included)
Specifications
Type Dispensing Bin
Material PETG
Color Clear
Shape Square
Mounting Type Tape Mount
Autoclavable Not Autoclavable