Learn More
Poltex Double Bulk Bin With Lid, 8 in. Wide Comp
SDP
Catalog No. 21377543
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Tape Mount
Wall Mount or Countertop
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-543 Wall Mount or Countertop
21-377-541 Magnetic Mount
21-377-542 Tape Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-377-543 Supplier Poltex Supplier No. BLKBNLDDBL8W
Add to Cart
Add to Cart
Simple 8 in. two-compartment bulk bin with top opening with lid for filling and front openings for dispensing.
Simple two-compartment bulk bin with lid
Durable and cleanable PETG construction
8 in. wide
Choose from:
  • Wall or countertop mount with rubber bumpons on base
  • VHB tape mount (very high bond tape included)
  • Magnet mount (powerful magnets included)
Specifications
Type Dispensing Bin
Material PETG
Color Clear
Shape Rectangular
Mounting Type Wall Mount or Countertop
Autoclavable Not Autoclavable